Skip to Content
Merck

Skip To

04-1664

Anti-Histidine Tag Antibody, clone RM146

clone RM146, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 02, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 12,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Clone:
RM146, monoclonal
Technique(s):
immunoprecipitation (IP): suitable, western blot: suitable
Application:
IP, WB
Citations:
-

CZK 12,800.00


Estimated to ship onJune 02, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

RM146, monoclonal

species reactivity (predicted by homology)

all

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HRG(3273)

General description

~52 and ~85 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Mixture of a peptide with 6xHis-Tag at the N-terminus and another peptide with 6xHis-Tag at the C-terminus.

Application

Immunoprecipitation Analysis (IP): 0.5-2 μg/mL from a representative lot detected Histidine Tag in HEK293T whole lysate control and IP by clone RM146.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
This Anti-Histidine Tag Antibody, clone RM146 is validated for use in Western Blotting, Immunoprecipitation for the detection of Histidine Tag.

Biochem/physiol Actions

This antibody reacts to recombinant proteins containing the 6xHis-Tag or 10xHis-Tag fused to either the amino or carboxy terminus. No cross reactivity with other endogenous protein in mammalian or bacteria cells.

Physical form

Format: Purified
Protein A purified
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Evaluated by Western Blotting in HEK293T cell lysate untransfected with His-Tag fusion protein X, transfected with His-Tag fusion protein X, E. coli lysate without His-Tag Protein Y, and E. coli lysate with His-Tag Protein Y.

Western Blotting Analysis (WB): 0.2 μg/mL of this antibody detected Histidine Tag in HEK293T cell lysate transfected with His-Tag fusion protein X and E. coli lysate with His-Tag Protein Y.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-53105-31105-949
clone

RM146, monoclonal

clone

4D11, monoclonal

clone

DG122-2A7, monoclonal

clone

HIS.H8, monoclonal

biological source

rabbit

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified antibody

antibody form

affinity purified immunoglobulin

antibody form

purified antibody

antibody form

purified antibody

Gene Information

human ... HRG(3273)

Gene Information

human ... HRG(3273)

Gene Information

-

Gene Information

-

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters