Skip to Content
Merck
All Photos(1)

Key Documents

Z111481

Sigma-Aldrich

Polystyrene vial for analog Wig-L-Bug® grinder/mixer

L 1 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

material

polystyrene vial

availability

not available in EU

technique(s)

IR spectroscopy: suitable

L

1 in.

capacity

1.0 mL

diam.

1/2 in.

Looking for similar products? Visit Product Comparison Guide

General description

With PE slip-on cap. For mulling with mineral oil. Use with Plexiglas® ball pestles, order separately.

Legal Information

Plexiglas is a registered trademark of Arkema
Wig-L-Bug is a registered trademark of Dentsply Rinn

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service