Key Documents
Z111538
Polystyrene vial for analog Wig-L-Bug® grinder/mixer
L 2 in.
About This Item
UNSPSC Code:
23151817
availability
not available in EU
L
2 in.
capacity
1.0 mL
diam.
1/2 in.
General description
With PE slip-on cap. For mulling with mineral oil. Use with Plexiglas® ball pestles, order separately.
Legal Information
Plexiglas is a registered trademark of Arkema
Wig-L-Bug is a registered trademark of Dentsply Rinn
Choose from one of the most recent versions:
Certificates of Analysis (COA)