WHA10334500
Whatman® prepleated qualitative filter paper for technical use, Grade 0860 1/2
L × W (100 mm) (300 mm), pack of 500 ea
Select a Size
CZK 2,930.00
Select a Size
About This Item
CZK 2,930.00
Recommended Products
packaging
pack of 500 ea
manufacturer/tradename
Cytiva
Whatman 10334500
parameter
60 sec/100 mL speed (Herzberg)
L × W
(100 mm) (300 mm)
thickness
170 μm
pore size
7-12 μm (Particle retention)
basis weight
75 g/m2
General description
- Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
- Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
- Increased total loading capacity as more filter area is available
- Maintained flow rate due to the reduction in filter paper contact with funnel side and, the self-supporting shape of the filter itself
- The prepleating does not significantly affect any of the technical data and the same figures may be used for the flat circles
Grade 0860 ½ : 7-12 μm (medium fast, smooth filter paper)
Comparable to grade 0858 but with a smooth surface, slightly thinner and faster.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)