Přejít k obsahu
Merck
  • Lidocaine alleviates morphine tolerance via AMPK-SOCS3-dependent neuroinflammation suppression in the spinal cord.
Lidocaine alleviates morphine tolerance via AMPK-SOCS3-dependent neuroinflammation suppression in the spinal cord.
Journal of neuroinflammation (2017-11-04)
Yan Zhang, Gao-Jian Tao, Liang Hu, Jie Qu, Yuan Han, Guangqin Zhang, Yanning Qian, Chun-Yi Jiang, Wen-Tao Liu
ANOTACE
Morphine tolerance is a clinical challenge, and its pathogenesis is closely related to the neuroinflammation mediated by Toll-like receptor 4 (TLR4). In Chinese pain clinic, lidocaine is combined with morphine to treat chronic pain. We found that lidocaine sufficiently inhibited neuroinflammation induced by morphine and improved analgesic tolerance on the basis of non-affecting pain threshold. CD-1 mice were utilized for tail-flick test to evaluate morphine tolerance. The microglial cell line BV-2 was utilized to investigate the mechanism of lidocaine. Neuroinflammation-related cytokines were measured by western blotting and real-time PCR. The level of suppressor of cytokine signaling 3 (SOCS3) and adenosine 5'-monophosphate (AMP)-activated protein kinase (AMPK)-related signaling pathway was evaluated by western blotting, real-time PCR, enzyme-linked immunosorbent assay (ELISA), and immunofluorescence staining. Lidocaine potentiated an anti-nociceptive effect of morphine and attenuated the chronic analgesic tolerance. Lidocaine suppressed morphine-induced activation of microglia and downregulated inflammatory cytokines, interleukin-1β (IL-1β), and tumor necrosis factor-alpha (TNF-α) via upregulating SOCS3 by activating AMPK. Lidocaine enhanced AMPK phosphorylation in a calcium-dependent protein kinase kinase β (CaMKKβ)-dependent manner. Furthermore, lidocaine decreased the phosphorylation of p38 mitogen-activated protein kinase (MAPK) and inhibited the nuclear factor-κB (NF-κB) in accordance with the inhibitory effects to TLR4. Lidocaine as a prevalent local anesthetic suppresses morphine tolerance efficiently. AMPK-dependent upregulation of SOCS3 by lidocaine plays a crucial role in the improvement of analgesic tolerance.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human SOCS3