- Enantioselective imidazole-directed allylation of aldimines and ketimines.
Enantioselective imidazole-directed allylation of aldimines and ketimines.
Organic letters (2007-08-10)
Nicholas R Perl, James L Leighton
PMID17685538
ANOTACE
A new chiral allylchlorosilane has been developed that allows the highly enantioselective allylation and crotylation of a range of 2-imidazolylaldimines and ketimines. The method may be exploited for the protecting group-free synthesis of a diverse array of imidazole-bearing chiral carbinamines and, when coupled with ring-closing metathesis reactions, allows the one-pot synthesis of unusual heterocyclic motifs with potential relevance in medicinal chemistry.
MATERIÁLY