Přejít k obsahu
Merck
Na+/H+ and Na+/NH+4 activities of zebrafish NHE3b expressed in Xenopus oocytes.
American journal of physiology. Regulatory, integrative and comparative physiology (2014-01-10)
Yusuke Ito, Akira Kato, Taku Hirata, Shigehisa Hirose, Michael F Romero
ANOTACE
Zebrafish Na(+)/H(+) exchanger 3b (zNHE3b) is highly expressed in the apical membrane of ionocytes where Na(+) is absorbed from ion-poor fresh water against a concentration gradient. Much in vivo data indicated that zNHE3b is involved in Na(+) absorption but not leakage. However, zNHE3b-mediated Na(+) absorption has not been thermodynamically explained, and zNHE3b activity has not been measured. To address this issue, we overexpressed zNHE3b in Xenopus oocytes and characterized its activity by electrophysiology. Exposure of zNHE3b oocytes to Na(+)-free media resulted in significant decrease in intracellular pH (pH(i)) and intracellular Na(+) activity (aNa(i)). aNa(i) increased significantly when the cytoplasm was acidified by media containing CO₂-HCO₃(-) or butyrate. Activity of zNHE3b was inhibited by amiloride or 5-ethylisopropyl amiloride (EIPA). Although the activity was accompanied by a large hyperpolarization of ∼50 mV, voltage-clamp experiments showed that Na(+)/H(+) exchange activity of zNHE3b is electroneutral. Exposure of zNHE3b oocytes to medium containing NH₃/NH₄(+) resulted in significant decreases in pH(i) and aNa(i) and significant increase in intracellular NH₄(+) activity, indicating that zNHE3b mediates the Na(+)/NH₄(+) exchange. In low-Na(+) (0.5 mM) media, zNHE3b oocytes maintained aNa(i) of 1.3 mM, and Na(+)-influx was observed when pHi was decreased by media containing CO₂-HCO₃(-) or butyrate. These results provide thermodynamic evidence that zNHE3b mediates Na(+) absorption from ion-poor fresh water by its Na(+)/H(+) and Na(+)/NH₄(+) exchange activities.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
USP
Amiloride hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride