Přejít k obsahu
Merck
  • Chromotropic character of bacterial acidic polysaccharides: Part II--Induction of metachromasy in cationic dye pinacyanol chloride by Klebsiella K10 capsular polysaccharide.
Chromotropic character of bacterial acidic polysaccharides: Part II--Induction of metachromasy in cationic dye pinacyanol chloride by Klebsiella K10 capsular polysaccharide.
Indian journal of biochemistry & biophysics (1989-04-01)
A Chakrabarti, R K Nath, A K Chakraborty
ANOTACE
The acidic capsular polysaccharide isolated from Klebsiella K10 exhibited chromotropic character with respect to induction of metachromasy in the cationic dye pinacyanol chloride (1-ethyl-2-[3-(1-ethyl-2(1H)-quinolylidene)propenyl]quinolinium chloride). Klebsiella K10 polymer consists of hexasaccharide repeating units containing one residue of glucuronic acid along with other neutral sugars in each repeating unit. It induces a metachromatic blue shift in the visible absorption spectrum of the dye from 600 nm to 500 nm. The spectral changes have been studied during interaction of the dye cations with the polyanions at different polymer/dye molar ratios. The polyanion-dye compounds are formed with polymer/dye stoichiometry of 1:1, indicating formation of stacking conformation. The complete reversal of polymer-induced metachromasy has also been observed by the addition of ethanol and urea.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Pinacyanol chloride
Sigma-Aldrich
1,1′-Diethyl-2,2′-carbocyanine iodide, 97%