Přejít k obsahu
Merck
  • The intestinal bioavailability of vaccenic acid and activation of peroxisome proliferator-activated receptor-α and -γ in a rodent model of dyslipidemia and the metabolic syndrome.
The intestinal bioavailability of vaccenic acid and activation of peroxisome proliferator-activated receptor-α and -γ in a rodent model of dyslipidemia and the metabolic syndrome.
Molecular nutrition & food research (2012-06-21)
Ye Wang, Maria M Jacome-Sosa, Megan R Ruth, Yan Lu, Jianheng Shen, Martin J Reaney, Shannon L Scott, Michael E R Dugan, Hope D Anderson, Catherine J Field, Spencer D Proctor, Donna F Vine
ANOTACE
Evidence suggests a neutral to beneficial role of certain trans fatty acids (TFA) from natural ruminant sources. Trans11-18:1 (vaccenic acid, VA), the most predominant ruminant TFA and a precursor to conjugated linoleic acid, has been shown to improve atherogenic dyslipidemia and symptoms of hepatic steatosis in animal models. The objective of this study was to assess the intestinal bioavailability of various VA sources including synthetic free fatty acid (FFA) and natural ruminant triglyceride forms, as well as the mechanistic pathways that mediate VA's bioactivity. VA acts as a partial agonist to both peroxisome proliferator-activated receptors (PPAR)-α and PPAR-γ in vitro, with similar affinity compared to commonly known PPAR agonists. It was further confirmed that VA at 30 and 100 μM concentrations suppressed cardiomyocyte hypertrophy vitro in a PPAR-α- and PPAR-γ-dependent manner. In vivo, feeding of VA (1%, w/w) resulted in increased mRNA and protein expression of PPAR-γ in the mucosa of JCR:LA-cp rats, a model of the metabolic syndrome (p < 0.01 and p < 0.05, respectively) compared to control. In addition, VA from a triglyceride source had greater intestinal bioavailability in vivo compared to VA provided in an FFA form (p < 0.01). The activation of PPAR-α- and PPAR-γ-dependent pathways provides a mechanistic explanation of how VA improves blood lipids and related metabolic disorders during conditions of hyperlipidemia. This report also supports the consideration of differential reporting of industrially produced versus natural TFA on food nutrient labels.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
trans-Vaccenic acid, ≥99% (capillary GC)