Přejít k obsahu
Merck
  • "Click chemistry" in tailor-made polymethacrylates bearing reactive furfuryl functionality: a new class of self-healing polymeric material.
"Click chemistry" in tailor-made polymethacrylates bearing reactive furfuryl functionality: a new class of self-healing polymeric material.
ACS applied materials & interfaces (2010-04-02)
A Amalin Kavitha, Nikhil K Singha
ANOTACE
This investigation reports the effective use of the Diels-Alder (DA) reaction, a "click reaction" in the preparation of thermally amendable and self-healing polymeric materials having reactive furfuryl functionality. In this case, the DA and retro-DA (rDA) reactions were carried out between the tailor-made homo- and copolymer of furfuryl methacrylate prepared by atom-transfer radical polymerization and a bismaleimide (BM). The kinetic studies of DA and rDA reactions were carried out using Fourier transform infrared spectroscopy. The DA polymers were insoluble in toluene at room temperature. When the DA polymers were heated at 100 degrees C in toluene, it was soluble. This is because of the cleavage between furfuryl functionality and BM. The chemical cross-link density was determined by the Flory-Rehner equation. The cross-linked polymer showed much greater adhesive strength at room temperature, but the adhesive strength was quite low at higher temperature. The self-healing capability was studied by using scanning electron microscopy analysis. The thermal and dynamic mechanical properties of the thermally amendable cross-linked materials were investigated by thermogravimetric analysis and dynamic mechanical analysis.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Furfuryl methacrylate, 97%, contains 200 ppm monomethyl ether hydroquinone as inhibitor