Přejít k obsahu
Merck
  • Ionic-liquid-promoted palladium-catalyzed multicomponent cyclocarbonylation of o-iodoanilines and allenes to form methylene-2,3-dihydro-1H-quinolin-4-ones.
Ionic-liquid-promoted palladium-catalyzed multicomponent cyclocarbonylation of o-iodoanilines and allenes to form methylene-2,3-dihydro-1H-quinolin-4-ones.
The Journal of organic chemistry (2007-04-06)
Fangguo Ye, Howard Alper
ANOTACE
The palladium-catalyzed cyclocarbonylation reaction of o-iodoanilines with allenes and CO in 1-butyl-3-methylimidazolium hexafluorophosphate afforded 3-methylene-2,3-dihydro-1H-quinolin-4-ones in moderate to excellent yields under a low pressure (5 atm) of CO. The ionic liquid, as the solvent and promoter, enhances the efficiency of the cyclocarbonylation reaction. The recyclability of the system of ionic liquid/catalyst/ligand was also demonstrated.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
3-Iodoaniline, 98%