Přejít k obsahu
Merck
  • Tridentate Directing Groups Stabilize 6-Membered Palladacycles in Catalytic Alkene Hydrofunctionalization.
Tridentate Directing Groups Stabilize 6-Membered Palladacycles in Catalytic Alkene Hydrofunctionalization.
Journal of the American Chemical Society (2017-10-04)
Miriam L O'Duill, Rei Matsuura, Yanyan Wang, Joshua L Turnbull, John A Gurak, De-Wei Gao, Gang Lu, Peng Liu, Keary M Engle
ANOTACE
Removable tridentate directing groups inspired by pincer ligands have been designed to stabilize otherwise kinetically and thermodynamically disfavored 6-membered alkyl palladacycle intermediates. This family of directing groups enables regioselective remote hydrocarbofunctionalization of several synthetically useful alkene-containing substrate classes, including 4-pentenoic acids, allylic alcohols, homoallyl amines, and bis-homoallylamines, under Pd(II) catalysis. In conjunction with previous findings, we demonstrate regiodivergent hydrofunctionalization of 3-butenoic acid derivatives to afford either Markovnikov or anti-Markovnikov addition products depending on directing group choice. Preliminary mechanistic and computational data are presented to support the proposed catalytic cycle.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
[2,2′-Bipyridine]-6-carboxylic acid hydrochloride