Merck
Hirschmann® EM dispenser, adjustable volume volume 0.4-2 mL, accuracy: 0.5%, precision: 0.1 CV%
Hirschmann® EM dispenser, adjustable volume
Hirschmann® EM dispenser, adjustable volume volume 0.4-2 mL, accuracy: 0.5%, precision: 0.1 CV%