Key Documents
58005
Supelco
Reagent sprayer for TLC plates
volume 250 mL, Bottle-Type
About This Item
UNSPSC Code:
41115711
technique(s)
thin layer chromatography (TLC): suitable
volume
250 mL
General description
The extraction phase or adsorbent phase can be coated on to the solid phase as a thin film. Coating preparation is of three types; dipping, brush painting and spraying. Spraying method was widely used due to its stability and reusability.
Application
Reagent sprayer may be used in thin-layer chromatography.
Compressed-air sprayers for spraying TLC plates. Sets include a glass-sprayer head and a choice of either bottle or tube with hexfoot stand.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
Je nám líto, ale pro tento produkt momentálně nemáme COA k dispozici online.
Zákazníci si také prohlíželi
Jae Hong Park et al.
The Laryngoscope, 130(2), 358-366 (2019-03-13)
In this study, we assessed the effectiveness of a tonsil-derived mesenchymal stem cell (TMSC)-transplanted polycaprolactone/beta-tricalcium phosphate prosthesis (specifically designed for easier fixing and grafting with a single scaffold) on rabbit mandible osteogenesis. The mandibles of 18 rabbits were exposed, and
Robert Farkaš et al.
Development, growth & differentiation, 58(6), 562-574 (2016-07-12)
The Drosophila salivary glands (SGs) were well known for the puffing patterns of their polytene chromosomes and so became a tissue of choice to study sequential gene activation by the steroid hormone ecdysone. One well-documented function of these glands is
Fatemeh S Mirnaghi et al.
Analytical chemistry, 83(15), 6018-6025 (2011-06-30)
Biocompatible C18-polyacrylonitrile (PAN) coating was used as the extraction phase for an automated 96-blade solid phase microextraction (SPME) system with thin-film geometry. Three different methods of coating preparation (dipping, brush painting, and spraying) were evaluated; the spraying method was optimum
Parameswaran Vijayakumar et al.
Marine biotechnology (New York, N.Y.), 22(3), 333-347 (2020-02-23)
Teleosts have the ability to regenerate their caudal fin upon amputation. A highly proliferative mass of undifferentiated cells called blastema forms beneath wound epidermis and differentiates to regenerate all missing parts of the fin. To date, the origin and fate
Sandra Loera-Serna et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(17), 4398-4411 (2019-01-05)
The metal-organic framework (MOF) HKUST-1 was employed as an interaction matrix for fundamental loading studies of anthraquinone dyes. Chosen dyes were alizarin (A), alizarin S (AS), disperse blue 1 (B1), disperse blue 3 (B3), disperse blue 56 (B56) and purpurin