Změnit zobrazení
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
6 pkg | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 13 000,00 Kč |
O této položce
UNSPSC Code:
12352200
NACRES:
NB.21
13 000,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlipackaging
pk of 6
Quality Segment
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
Equip alternative valves in your 12-port or 24-port standard Visiprep vacuum manifold with these long stem flow control valves if you intend to use all ports of the manifold with 12, 20, or 60 mL SPE tubes.
Note: Not for use with Visiprep DL (Disposable Liner) models
Note: Not for use with Visiprep DL (Disposable Liner) models
Equip alternative valves in your 12-port or 24-port standard Visiprep™ vacuum manifold with these long stem flow control valves if you intend to use all ports of the manifold with 12, 20, or 60 mL SPE tubes.
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
1 of 1
Tato položka | |||
|---|---|---|---|
| description pk of 6 | description for use with DL (Disposable Liner) 12- and 24-port models, pk of 2 | description for use with standard 12- and 24-port models, pk of 2 | description pk of 12 |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable |
| packaging pk of 6 | packaging pk of 2 | packaging pk of 2 | packaging pk of 12 |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?