Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
960 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 5 500,00 Kč |
O této položce
5 500,00 Kč
material
colorless , colorless tip, polypropylene (Virgin)
sterility
sterile
feature
filter, DNase free, RNase free, barrier
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sartorius 791001F
maximum volume
1 mL
volume range
50-1000 μL
volume
50—1000 μL
binding type
low binding surface
General description
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
- Pipette foaming liquids like buffers and proteins
- Pipette solvents
- Reverse pipetting
- Multiple dispensing functions of electronic pipettes
Other Notes
1 of 1
Tato položka | |||
|---|---|---|---|
| description volume range 50-1000 μL, sterile, pack of 960 ea (10 racks of 96 tips) | description volume range 50-1200 μL, sterile, pack of 960 ea (10 racks of 96 tips) | description volume range 2-120 μL, sterile, pack of 960 ea (10 racks of 96 tips) | description ART 20, volume range 0.5-20 μL, sterile |
| feature barrier, RNase free, DNase free, filter | feature barrier, DNase free, filter, RNase free | feature filter, barrier | feature RNase free, barrier, DNase free |
| volume 50—1000 μL | volume 50—1250 μL | volume 2—120 μL | volume 0.5—20 μL |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Sartorius 791001F | manufacturer/tradename Sartorius 791211F | manufacturer/tradename Sartorius 790101F | manufacturer/tradename Molecular BioProducts 2149 |
| material polypropylene (Virgin), colorless tip | material colorless , polypropylene (Virgin), colorless tip | material colorless , polypropylene (Virgin) | material colorless tip |
| binding type low binding surface | binding type low binding surface | binding type low binding surface | binding type - |
Still not finding the right product?
Explore all of our products under Sartorius filter pipette tips
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?