Z743999
Worthington Accessories
necktube core for LS6000 liquid nitrogen refrigerator
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Doporučené produkty
packaging
pack of 1 ea
manufacturer/tradename
Worthington RS60-9C16
technique(s)
cryopreservation: suitable
General description
All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)