Key Documents
Z742710
BenchMixer XLQ QuEChERs Shaker/Vortexer, tube rack for 7 x 50 mL tubes, PP
About This Item
UNSPSC Code:
41103816
NACRES:
NB.73
material
polypropylene
product line
BenchMixer
technique(s)
QuEChERS: suitable
General description
Foam tube horizontal tube rack to hold 7x50ml tubes for QuEChERs applications in Benchmixer XLQ shaker/vortexer.
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.