Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1000 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 160,00 Kč |
O této položce
2 160,00 Kč
material
conical bottom bottom, polypropylene tube, straight neck, translucent tube
sterility
non-sterile
feature
RCF 3,000 × g, neck thread push fit
packaging
pack of 1000 ea
manufacturer/tradename
Simport T400-3A
capacity
5 mL
size
12 mm × 75 mm
General description
1 of 1
Tato položka | |||
|---|---|---|---|
| description polypropylene tube | description polystyrene tube | description - | description - |
| sterility non-sterile | sterility non-sterile | sterility - | sterility - |
| manufacturer/tradename Simport T400-3A | manufacturer/tradename Simport T400-3 | manufacturer/tradename 231721986 | manufacturer/tradename 231752155 |
| material conical bottom bottom, straight neck, polypropylene tube, translucent tube | material polystyrene tube, round bottom, straight neck, translucent tube | material soda-lime glass vial | material polypropylene screw cap, soda-lime glass vial |
| feature RCF 3,000 × g, neck thread push fit | feature RCF 1,400 × g, neck thread push fit | feature - | feature seal (rubber) |
| capacity 5 mL | capacity 5 mL | capacity - | capacity - |
| size 12 mm × 75 mm | size 12 mm × 75 mm | size - | size - |
Still not finding the right product?
Explore all of our products under Disposable culture tubes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?