O této položce
NACRES:
NB.22
UNSPSC Code:
41121706
eCl@ss:
32040302
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
polystyrene tube, round bottom, straight neck, translucent tube
sterility
non-sterile
feature
RCF 1,400 × g, neck thread push fit
packaging
pack of 1000 ea
manufacturer/tradename
Simport T400-3
technique(s)
cell culture: suitable
capacity
5 mL
size
12 mm × 75 mm
General description
Ideal for use in bacteriology, RIA, coagulation, and other routine laboratory procedures. Polypropylene tubes can be centrifuged at 3000 × g and sterilized at 120°C. Polystyrene tubes can be centrifuged at 1400 × g, but can not be autoclaved.
Still not finding the right product?
Explore all of our products under Disposable culture tubes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?