Key Documents
Z328499
BRAND® Transferpettor caps
maximum volume 25 μL
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
glass
sterility
non-sterile
feature
color-coded white (x2)
DE-M marking
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701906
BRAND
maximum volume
25 μL
Other Notes
BR701906 (India)