Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 120,00 Kč |
O této položce
2 120,00 Kč
material
clear borosilicate glass 3.3 , round bottle
joint
joint 29/32 standard ground joint, joint ST/NS neck29/32
feature
without cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 23310
capacity
2 L
diam. × H
160 mm × 200 mm
General description
Other Notes
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description capacity 2 L, clear borosilicate glass 3.3 | description capacity 2 L, amber borosilicate glass 3.3 | description capacity 2 L, clear soda-lime glass | description volume 1000 mL, low-density polyethylene |
| manufacturer/tradename BRAND 23310 | manufacturer/tradename BRAND 23320 | manufacturer/tradename BRAND 126965 | manufacturer/tradename BRAND 129060 |
| feature without cap | feature without cap | feature without cap | feature cap |
| material clear borosilicate glass 3.3 , round bottle | material amber borosilicate glass 3.3 , round bottle | material clear soda-lime glass , round bottle | material low-density polyethylene , narrow-mouth bottle, round bottle, translucent bottle |
| capacity 2 L | capacity 2 L | capacity 2 L | capacity 1000 mL |
| joint joint 29/32 standard ground joint, joint ST/NS neck29/32 | joint joint 29/32 standard ground joint, joint ST/NS neck29/32 | joint joint 29/32 standard ground joint, joint ST/NS neck29/32 | joint - |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging - |
Still not finding the right product?
Explore all of our products under BRAND® reservoir bottle for automatic burettes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?