Skip To
Z324019
BRAND® reservoir bottle for automatic burettes
capacity 2 L, clear soda-lime glass
Synonym(s):
2000mL burette bottle, burette reservoir bottle, clear laboratory bottle, glass bottle, reservoir bottle
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$44.20
About This Item
NACRES:
NB.43
UNSPSC Code:
41121800
$44.20
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear soda-lime glass , round bottle
joint
joint 29/32 standard ground joint, joint ST/NS neck29/32
feature
without cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 126965
capacity
2 L
diam. × H
160 mm × 200 mm
General description
DURAN® spare reservoir bottles for automatic burettes.
Other Notes
BR126965 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
DURAN is a registered trademark of DWK Life Sciences
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
Z323985Z323977Z323993
manufacturer/tradename
BRAND 126965
manufacturer/tradename
BRAND 127065
manufacturer/tradename
BRAND 23320
manufacturer/tradename
BRAND 23310
material
clear soda-lime glass , round bottle
material
amber soda-lime glass , round bottle
material
amber borosilicate glass 3.3 , round bottle
material
clear borosilicate glass 3.3 , round bottle
feature
without cap
feature
without cap
feature
without cap
feature
without cap
capacity
2 L
capacity
2 L
capacity
2 L
capacity
2 L
joint
joint 29/32 standard ground joint, joint ST/NS neck29/32
joint
joint 29/32 standard ground joint, joint ST/NS neck29/32
joint
joint 29/32 standard ground joint, joint ST/NS neck29/32
joint
joint 29/32 standard ground joint, joint ST/NS neck29/32
diam. × H
160 mm × 200 mm
diam. × H
160 mm × 200 mm
diam. × H
160 mm × 200 mm
diam. × H
160 mm × 200 mm
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)