WPB01547WA
Whirl-Pak® Filter Bag Without Tape/Wire
1627 mL capacity(55 oz), pack of 250 ea
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Doporučené produkty
packaging
pack of 250 ea
manufacturer/tradename
B01547WA (Nasco)
W × L × thickness
19 cm × 30 cm × 0.102 mm
capacity
1627 mL capacity(55 oz)
General description
Filter blender bag without tape and wire closure.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)