Key Documents
MTCA8803
MTC Bio BluBlot HS Autoradiography Flim
W × L 5 in. × 7 in.
Synonyma:
blotting film, western imaging film
About This Item
UNSPSC Code:
41105341
NACRES:
NB.15
packaging
pack of 100 ea
manufacturer/tradename
MTC Bio A8803
W × L
5 in. × 7 in.
General description
BluBlot HS Autoradiography film, 5x7in, 100 sheets/box
Features and Benefits
  • High Contrast
  • Reduced Background
  • Publication Quality
  • Suitable for 14C, 32P, 33P, 125I, and 35S
  • Double Emulsion
Legal Information
BluBlot is a trademark of MTC Bio
MTC is a trademark of MTC Bio