HS120114
Mega Rack® tube rack
double, for 13-16 mm tubes, white
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
Doporučené produkty
material
polypropylene
feature
Holds 240 x 10 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120114
size
673x267x90 mm
color
white
General description
Mega Rack High-capacity storage rack for sample and blood collection tubes.
- Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
- Sturdy and light weight material for easy transport
- Alphanumeric identification for quick location and retrieval of tubes
- Place different test groupings into one rack and eliminate need for multiple tube racks
- Autoclavable
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)