Hlavní dokumenty
GE80-1129-46
IEF sample application piece, for MultiPhor II
pack of 200 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
packaging
pack of 200 ea
manufacturer/tradename
Cytiva 80112946
General description
Samples can be applied on the surface of the gels either by pipetting on pieces from cellulose-cotton, which are placed on the gel surface at the optimal pH location, or on to a silicone applicator strip with sample holes, laid on the gel surface.
Legal Information
MultiPhor is a trademark of Cytiva
Storage Class
13 - Non Combustible Solids
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)