Key Documents
GE80-1106-88
Glass Plates
U-frame, H × W × D 200 mm × 260 mm × 1 mm, pack of 2 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
packaging
pack of 2 ea
manufacturer/tradename
Cytiva 80110688
H × W × D
200 mm × 260 mm × 1 mm
Související kategorie
Storage Class