Key Documents
GE27-4732-01
Sigma-Aldrich
Poly(I)·Poly(C)
About This Item
UNSPSC Code:
41106305
NACRES:
NA.51
General description
Since it was discovered that the double-stranded polyribonucleotide poly(I)·poly(C) induces interferon in rabbit cells, there has been much interest in its effectiveness for inducing human interferon in vitro.
Features and Benefits
For stimulation of interferon production in vitro
Supplied as a sterile lyophilized powder
Storage Class