Key Documents
EP5910769008
Eppendorf® Adapter
for 50 mL conical tubes, for rotor S-4xUniv., pkg of 2 pieces
About This Item
UNSPSC Code:
41103911
NACRES:
NB.78
feature
for 50 mL conical tubes
for rotor S-4xUniv.
packaging
pack of 2 ea
pkg of 2 pieces
manufacturer/tradename
Eppendorf® 5910769008
technique(s)
centrifugation: suitable
compatibility
for use with Universal bucket
Legal Information
Eppendorf is a registered trademark of Eppendorf AG