Hlavní dokumenty
EP5895160000
Rotor for Eppendorf® 5910 R Centrifuge
F-48x15, incl. 48 steel sleeves and adapters, for 40x conical 15 mL tubes, pack of 1 ea
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 40 x 15 mL conical tubes
F-48x15, incl. 48 steel sleeves and adapters, for 40x conical 15 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895160000
parameter
5,500 max. RCF (5,005 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5910R
General description
Rotor for Eppendorf(R)5910R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)