Key Documents
EP5427755004
Rotor for Eppendorf® 5430/5430R Centrifuges
F-45-48-11, non aerosol tight, to hold, 48 x 1.5/2.0 mL tubes, lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
lid
to hold 48 x 1.5/2.0 mL tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427755004 (Global)
parameter
12,700 rpm
18,210 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG