Key Documents
EP4966506003
Telescoping aspirating tube for Eppendorf® Varispenser® 2 / 2x
adjustable, L 250-480 mm, for 2/5/10 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.01
feature
adjustable
for 2/5/10 mL
manufacturer/tradename
Eppendorf® 4966506003
L
250-480 mm
Související kategorie
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Varispenser is a registered trademark of Eppendorf AG