Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 4 900,00 Kč |
O této položce
4 900,00 Kč
material
gray
feature
single-channel, mechanical
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 3121000031 (Global)
volume
20 μL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description single-channel, volume 20 μL, gray | description single-channel, volume 20 μL, yellow | description single-channel, volume 10 μL, yellow | description single-channel, volume 25 μL, yellow |
| feature single-channel, mechanical | feature mechanical, single-channel | feature single-channel, mechanical | feature single-channel, mechanical |
| manufacturer/tradename Eppendorf® 3121000031 (Global) | manufacturer/tradename Eppendorf® 3121000040 (Global) | manufacturer/tradename Eppendorf® 3121000023 (Global) | manufacturer/tradename Eppendorf® 3121000058 (Global) |
| volume 20 μL | volume 20 μL | volume 10 μL | volume 25 μL |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| material gray | material yellow | material yellow | material yellow |
Still not finding the right product?
Explore all of our products under Eppendorf Research® plus Pipette, fixed volume
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?