Hlavní dokumenty
EP0030622103
Eppendorf® Microplate 384/F-PP, 384 wells
white polypropylene, flat bottom clear wells, sterile, pkg of 80 ea
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material
flat bottom clear wells
white polypropylene
sterility
sterile; autoclaved
sterile
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030622103
technique(s)
PCR: suitable
size
384 wells
General description
Experience a new level of transparency with Eppendorf Microplates. With the highest quality polypropylene and superior well design, you can pipette, control and recover samples with minimal effort.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG