Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
200 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 360,00 Kč |
O této položce
2 360,00 Kč
sterility
non-sterile
feature
DNA/RNA LoBind, PCR clean
packaging
pkg of 200 tubes (4 bags of 50 tubes)
manufacturer/tradename
Eppendorf® 0030122232
measuring range
50 mL
technique(s)
PCR: suitable
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Eppendorf® 0030122232 | manufacturer/tradename Eppendorf® 0030108302 | manufacturer/tradename Eppendorf® 0030122208 | manufacturer/tradename Eppendorf® 0030122216 |
| packaging pkg of 200 tubes (4 bags of 50 tubes) | packaging pkg of 100 tubes (2 bags of 50 tubes) | packaging pkg of 200 tubes (4 bags of 50 tubes) | packaging pack of 200 ea |
| feature DNA/RNA LoBind, PCR clean | feature protein | feature PCR clean, DNA/RNA LoBind | feature PCR clean, graduations, neck thread External thread |
| technique(s) PCR: suitable | technique(s) - | technique(s) PCR: suitable | technique(s) PCR: suitable |
| measuring range 50 mL | measuring range 5.0 mL | measuring range 15 mL | measuring range - |
Still not finding the right product?
Explore all of our products under Eppendorf® LoBind microcentrifuge tubes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?