Hlavní dokumenty
EP0030076249
Eppendorf® Empty Box 2.0 for epT.I.P.S.® Reloads
new
for tips 10 – 300 µL
About This Item
UNSPSC Code:
41121600
feature
for tips 10 – 300 µL
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 0030076249
General description
epT.I.P.S.® Box 2.0 for epT.I.P.S.® Reloads, empty box, does not include tips, Eppendorf Quality, 10 – 300 µL
application
General liquid handling
Features and Benefits
  • epT.I.P.S.® Box 2.0 – empty reusable boxes for tip volumes of 10 µL - 5 mL
  • epT.I.P.S.® Box 2.0 – all three sizes are availabe in Eppendorf Quality
  • Autoclavable at 121 °C (20 min.) for subsequent use
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)