Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|
O této položce
material
flat bottom flask, polycarbonate , polypropylene cap, vent cap, wide-mouth , wide-mouth neck
sterility
sterile
feature
cap, closure type Vented cap
packaging
pack of 1 ea, case of 4 ea
manufacturer/tradename
Corning 431684
capacity
5 L
Features and Benefits
- Corning® Erlenmeyer flasks are ideal for shaker culture applications, and storage.
- Baffled bottom flasks are designed to scale up suspension culture efficiently.
- Molded-in graduations for accurate measurements.
- A large vent cap allows gas exchange and reduces spilling.
- Highest sterility assurance level (SAL) of 10-6
- RNase-/DNase-free and nonpyrogenic.
- Flasks are autoclavable once at 121°C, 15psi for 20 minutes. Repeated autoclaving weakens the flask and is not recommended. Vent cap is not autoclavable.
Other Notes
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description baffled bottom, polycarbonate, sterile | description plain bottom, with aseptic connections, polycarbonate, sterile, case of 2 ea | description baffled bottom, with aseptic connections, polycarbonate, sterile, case of 2 ea | description 2L Erlenmeyer Flask w/ Vent Cap, flat bottom, tubing I.D. 1/8 in., polycarbonate, sterile, 3/cs |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Corning 431684 | manufacturer/tradename Corning 11501 | manufacturer/tradename Corning 11502 | manufacturer/tradename Corning 431512 |
| material polypropylene cap, vent cap, wide-mouth neck, flat bottom flask, polycarbonate | material polypropylene cap, vent cap, polycarbonate , plain bottom, wide-mouth neck | material flat bottom flask, wide-mouth neck, polycarbonate , vent cap, polypropylene cap | material orange polypropylene cap, polycarbonate (medical grade), wide-mouth , flat bottom flask, polyethersulfone membrane, wide-mouth neck |
| capacity 5 L | capacity - | capacity - | capacity 2 L |
| feature cap, closure type Vented cap | feature cap | feature cap | feature closure type Vented cap |
| packaging pack of 1 ea, case of 4 ea | packaging case of 2 ea | packaging case of 2 ea | packaging case of 3, pack of 1 |
Still not finding the right product?
Explore all of our products under Corning® Erlenmeyer flask, 5 L
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?