Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
20 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 9 040,00 Kč |
O této položce
NACRES:
NB.13
UNSPSC Code:
41121812
Material:
polystyrene , square
Sterility:
sterile; γ-irradiated
Manufacturer/tradename:
Corning 431301
9 040,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
polystyrene , square
sterility
sterile; γ-irradiated
manufacturer/tradename
Corning 431301
H
12.5 mm
L × W
245 mm × 245 mm
surface area
500 cm^2
General description
- Corning® square bioassay dishes are made from optically clear polystyrene.
- Stacking bead design facilitate secure stacking and prevents slipping.
- Compatible with automated colony picking instruments.
- Certified nonpyrogenic dishes are supplied with lids.
Legal Information
Corning is a registered trademark of Corning, Inc.
1 of 1
Tato položka | |||
|---|---|---|---|
| description low profile, without handles, not TC treated, square | description without handles, TC treated, square | description D × H 60 mm × 15 mm | description D × H 150 mm × 25 mm |
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile | sterility sterile |
| manufacturer/tradename Corning 431301 | manufacturer/tradename Corning 431110 | manufacturer/tradename Corning 430589 | manufacturer/tradename Corning 430597 |
| material square , polystyrene | material square , polystyrene | material round , polystyrene | material polystyrene , round |
| L × W 245 mm × 245 mm | L × W 245 mm × 245 mm | L × W - | L × W - |
| surface area 500 cm^2 | surface area 500 cm2 | surface area 21 cm2 | surface area 148 cm2 |
| H 12.5 mm | H 25 mm | H - | H - |
Still not finding the right product?
Explore all of our products under Corning® square bioassay dishes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?