Key Documents
CLS353910
Corning® Falcon® 96 Well Cell Culture Plate
round clear polystyrene, lid: no, non-sterile
About This Item
UNSPSC Code:
41122102
NACRES:
NB.13
material
round clear polystyrene
sterility
non-sterile
feature
lid: no
manufacturer/tradename
Corning 353910
technique(s)
cell culture | mammalian: suitable
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.