Key Documents
CLS11415
Corning® Erlenmeyer flask, with aseptic connector
volume 250 mL, I.D. 1/8 in., sterile
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
material
flat bottom flask
polycarbonate
polypropylene cap
description
plain
sterility
sterile
feature
cap: no
C-Flex tubing
packaging
case of 4
manufacturer/tradename
Corning 11415
I.D.
1/8 in.
capacity
250 mL
volume
250 mL
fitting
male luer lock