Hlavní dokumenty
BMSMW9600WSHB
Sigma-Aldrich
Extra Wash Bottle Assembly for SmartWasher™ microplate Washer
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
manufacturer/tradename
Benchmark Scientific MW9600-WSHB
General description
Chemically resistant wash bottle assembly for SmartWasher microplate Washer with includes level detection
Application
ELISA assays
Features and Benefits
  • Chemically resistant
  • Level detection
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)