Key Documents
Z803200
Hellma® cuvettes for light-scattering measurements
High Performance Quartz Glass, I.D. 8 mm, spectral range 200-2500 nm, chamber volume 2800 μL
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
material
Suprasil® quartz
manufacturer/tradename
Hellma 540.110-QS
parameter
200-2500 nm spectral range
technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
I.D.
8 mm
O.D.
75x10 mm
chamber volume
2800 μL
General description
with PTFE stoppers
Packaging
Sold as packages containing a single cell (1EA = 1 cell)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
related product
Č. produktu
Popis
Stanovení ceny
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)