O této položce
NACRES:
JA.13
UNSPSC Code:
41104200
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlipackaging
pkg of 1 unit
compatibility
for use with SDS
General description
This Automatic Sanitization Module (ASM) has an integrated UV lamp to prevent bacterial and biofilm growth in stored pure water and on tank inner surfaces.
For use with the SDS 500 pure water storage and distribution system.
For use with the SDS 500 pure water storage and distribution system.
Application
The ASM protects stored pure water from bacterial proliferation and biofilm formation.
Features and Benefits
- 254 nm UV lamp for germicidal effect.
Disclaimer
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Still not finding the right product?
Explore all of our products under Automatic Sanitization Module (ASM)
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| TANKSDS2ASM | 04054839077067 |