O této položce
NACRES:
JA.13
UNSPSC Code:
41104200
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlipackaging
pkg of 1 unit
compatibility
for use with AFS®, for use with Elix® Essential UV, for use with Elix® Essential, for use with RiOs™ Essential
General description
The mounting bracket easily installs a water purification unit high up on a wall, leaving benchtops free of clutter for optimized lab space and a more efficient working environment.
This bracket is compatible with RiOs™ Essential and Elix® Essential water purification Systems.
This bracket is compatible with RiOs™ Essential and Elix® Essential water purification Systems.
Application
Save lab bench space.
Features and Benefits
- Easy to install accessory.
- Saves lab bench space.
Legal Information
AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Wall Mounting Bracket
Globální číslo obchodní položky
| Skladová položka | GTIN |
|---|---|
| SYSTFIX01 | 04053252725791 |