Hlavní dokumenty
NA38/22TR
Millipore
EPDM Diaphragm for NovAseptic® Valve
ethylene propylene diene monomer rubber
About This Item
UNSPSC Code:
31411800
Quality Level
product line
NovAseptic®
parameter
-1-6 bar pressure range (-14,5-87 psi) (gramm)
-30-135 °C temp. range (-22-275 °F)
Application
The NovAseptic® Valve is specifically designed for aseptic applications and complies with the most stringent cleanability and sterilization requirements. The focus on aseptic design is a significant feature for all valves. The EPDM diaphragms are used in NovAseptic® Valves where ever compatible fluid application is concerned.
Features and Benefits
NovAseptic® valves are used within a range of biopharm unit operations and applications. These include valves for tank outlets, directional process flow control, divert valves for filter housings as well as valve assemblies for high value unit operations such as membrane and chromatography processes. The aseptic design of the NovAseptic® valve makes the process easier to validate, with diaphragm materials such as, EPDM, Silicone and PTFE available. These materials fulfill FDA criteria and are USP Class VI tested. NovAseptic® Valves are specifically designed to comply with the most stringent cleanability and sterility requirements
Packaging
The Diaphragm is packaged in a closed box
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany