Hlavní dokumenty
96100250
Millipore
Vantage L Laboratory Columns
Vantage L Laboratory Column VL 11 x 250
Synonyma:
Vantage L Laboratory Column VL 11 x 250
About This Item
UNSPSC Code:
23151816
eCl@ss:
32011301
Quality Level
product line
Vantage
parameter
operating temperature at 2-40 °C
6.9 bar max. inlet pressure (100 psi)
7 bar max. pressure (100 psid)
technique(s)
protein purification: suitable
L
250 mm
area
1 cm2 , cross-sectional
bed H
0-250 mm
bed volume
0-25 mL
diam.
11.5-11.6 mm
Legal Information
VANTAGE is a trademark of Merck KGaA, Darmstadt, Germany