Z740625
Aldrich® Essentials crystallizing dish, without spout
capacity 45 mL, dish diam. × H 40 mm × 25 mm
About This Item
UNSPSC Code:
41121812
NACRES:
NB.43
Doporučené produkty
material
borosilicate glass
packaging
pack of 6 ea
capacity
45 mL
dish diam. × H
40 mm × 25 mm
Související kategorie
General description
Clear borosilicate glass crystalizing dishes, with pour spout for easy dispensing of liquids. Not graduated.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)