Key Documents
Z681768
MyFuge 12 Mini Centrifuge Adapters
to hold, 0.2 mL tubes
About This Item
UNSPSC Code:
41103911
NACRES:
NB.78
feature
to hold 0.2 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
Benchmark Scientific C1008-A2-6
Benchmark Scientific
technique(s)
centrifugation: suitable
compatibility
for use with MC-12 High Speed Microcentrifuge
for use with MC-24 High Speed Microcentrifuge
for use with myFuge 12 Mini Centrifuge
for use with myFuge 5 MicroCentrifuge
for use with myFuge Mini Centrifuge
General description
Pack of 6 adapters to use 0.2mL tubes in Benchmark MyFuge, MC-24, and MC-12 centrifuges
Legal Information
MyFuge is a trademark of Benchmark Scientific, Inc.