Hlavní dokumenty
Z679283
Microgard 2500 disposable oversleeves, one size
L × W 400 mm × 200 mm, CE compliant
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
feature
CE compliant
L × W
400 mm × 200 mm
General description
Protects arms and sleeves.
  • Made from Microgard, a microporous PP laminate
  • Stiched seams
  • Integral elastication at both ends
Legal Information
Microgard is a trademark of Microgard Limited
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)